4-Benzyloxy-3-methoxyphenylacetic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

B4800-1G B4800-5G
TCIAB4800-1GEA 88 EUR
TCIAB4800-1G TCIAB4800-5G
4-Benzyloxy-3-methoxyphenylacetic acid ≥98.0% (by GC, titration analysis)
4-Benzyloxy-3-methoxyphenylacetic acid
Formula: C₁₆H₁₆O₄
MW: 272,30 g/mol
Número MDL: MFCD00185562
Núm. CAS: 29973-91-9

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR