N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: N-benzyloxycarbonyl-3-fluoro-4-morpholinoaniline, (3-Fluoro-4-morpholinophenyl)carbamic acid Benzyl ester, Benzyl (3-Fluoro-4-morpholinophenyl)carbamate

B4810-5G B4810-25G
TCIAB4810-5GEA 32 EUR
TCIAB4810-5G TCIAB4810-25G
N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline ≥98.0% (by HPLC, total nitrogen)
N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline
Formula: C₁₈H₁₉FN₂O₃
MW: 330,359 g/mol
Número MDL: MFCD06658240
Núm. CAS: 168828-81-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR