(1E,4E)-1,5-Bis[3,5-bis(methoxymethoxy)phenyl]-1,4-pentadiene-3-one ≥97.0% (by HPLC)
Proveedor: TCI
Sinónimos:
GO-Y030
B4823-20MG
B4823-100MG
TCIAB4823-20MGEA
66
EUR
TCIAB4823-20MG
TCIAB4823-100MG
(1E,4E)-1,5-Bis[3,5-bis(methoxymethoxy)phenyl]-1,4-pentadiene-3-one ≥97.0% (by HPLC)
(1E,4E)-1,5-Bis[3,5-bis(methoxymethoxy)phenyl]-1,4-pentadiene-3-one
Formula:
C₂₅H₃₀O₉ MW: 474,51 g/mol |
Núm. CAS:
917813-62-8 |
Resultados test especificaciones
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Melting point | 90.0 to 94.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...