1-Butyl-1-methylpiperidinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-butyl-N-methyl-piperidinium bis((trifluoromethyl)sulphonyl)imide
B4844-5G
B4844-25G
TCIAB4844-5GEA
41
EUR
TCIAB4844-5G
TCIAB4844-25G
1-Butyl-1-methylpiperidinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
1-Butyl-1-methylpiperidinium bis(trifluoromethanesulphonyl)imide
Formula:
C₁₂H₂₂F₆N₂O₄S₂ MW: 436,43 g/mol |
Número MDL:
MFCD14156017 Núm. CAS: 623580-02-9 |
Resultados test especificaciones
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...