(S)-1-Boc-2-methylpiperazine ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: (S)-2-Methyl-1-piperazinecarboxylic acid tert-Butyl ester, tert-Butyl (S)-2-Methyl-1-piperazinecarboxylate, (S)-tert-Butyl 2-methylpiperazine-1-carboxylate, (S)-1-Boc-2-methylpiperazine, (S)-1-(tert-Butoxycarbonyl)-2-methylpiperazine

B4846-1G B4846-5G
TCIAB4846-1GEA 38 EUR
TCIAB4846-1G TCIAB4846-5G
(S)-1-Boc-2-methylpiperazine ≥98.0% (by GC, titration analysis)
(S)-1-Boc-2-methylpiperazine
Formula: C₁₀H₂₀N₂O₂
MW: 200,2780 g/mol
Número MDL: MFCD01862121
Núm. CAS: 169447-70-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 43.0 to 47.0 deg-C
Specific rotation [a]20/D +68.0 to +72.0 deg (C=0.65, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR