Boc-Phe(4-CN)-OH ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Boc-4-cyano-Phe-OH, N-Boc-4-cyano-L-phenylalanine, Boc-Phe(4-CN)-OH, N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine
B4877-5G
B4877-1G
TCIAB4877-5GEA
177
EUR
TCIAB4877-5G
TCIAB4877-1G
Boc-Phe(4-CN)-OH ≥98.0% (by HPLC, titration analysis)
Boc-Phe(4-CN)-OH
Formula:
C₁₅H₁₈N₂O₄ MW: 290.319 g/mol |
Número MDL:
MFCD00672526 Núm. CAS: 131724-45-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -27.0 to -35.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...