4-(4-Bromofenil)morfolina ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

B4890-5G B4890-25G
TCIAB4890-5GEA 101 EUR
TCIAB4890-5G TCIAB4890-25G
4-(4-Bromofenil)morfolina ≥98.0% (by GC, titration analysis)
4-(4-Bromofenil)morfolina
Formula: C₁₀H₁₂BrNO
MW: 242,12 g/mol
Número MDL: MFCD04112483
Núm. CAS: 30483-75-1

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR