4-[(4-Bromophenyl)(p-tolyl)amino]benzaldehyde ≥95.0% (por GC)

Proveedor: TCI

B4922-200MG B4922-1G
TCIAB4922-200MGEA 97 EUR
TCIAB4922-200MG TCIAB4922-1G
4-[(4-Bromophenyl)(p-tolyl)amino]benzaldehyde ≥95.0% (por GC)
4-[(4-Bromophenyl)(p-tolyl)amino]benzaldehyde
Formula: C₂₀H₁₆BrNO
Número MDL: MFCD16658914
Núm. CAS: 733744-98-4

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 115.0 to 119.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR