2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: (2-Butylbenzofuran-3-yl)(4-hydroxy-3,5-diiodophenyl)methanone, L-3373, (2-Butyl-3-benzofuranyl)(4-hydroxy-3,5-diiodophenyl)methanone

B4938-1G B4938-5G
TCIAB4938-1GEA 78 EUR
TCIAB4938-1G TCIAB4938-5G
2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran ≥98.0% (by HPLC, titration analysis)
2-Butyl-3-(4-hydroxy-3,5-diiodobenzoyl)benzofuran
Formula: C₁₉H₁₆I₂O₃
MW: 546,14 g/mol
Número MDL: MFCD02675787
Núm. CAS: 1951-26-4
EINECS: 217-773-7

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 143.0 to 147.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR