2-(4-Bromophenyl)triphenylene ≥98.0% (por GC)

Proveedor: TCI

B4955-1G B4955-5G
TCIAB4955-1GEA 98 EUR
TCIAB4955-1G TCIAB4955-5G
2-(4-Bromophenyl)triphenylene ≥98.0% (por GC)
2-(4-Bromophenyl)triphenylene
Formula: C₂₄H₁₅Br
MW: 383,29 g/mol
Núm. CAS: 1158227-56-5

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR