Boc-D-Chg-OH ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Boc-D-Chg-OH, N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine, N-Boc-2-cyclohexyl-D-glycine, Boc-cyclohexyl-D-Gly-OH, (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid, BOC-D-α-cyclohexylglycine, N-Boc-D-2-cyclohexylglycine
B4975-1G
B4975-5G
TCIAB4975-1GEA
17
EUR
TCIAB4975-1G
TCIAB4975-5G
Boc-D-Chg-OH ≥98.0% (by HPLC, titration analysis)
Boc-D-Chg-OH
Formula:
C₁₃H₂₃NO₄ MW: 257.33 g/mol Pto. de fusión: 75°C |
Número MDL:
MFCD00133629 Núm. CAS: 70491-05-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -12.0 to -9.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...