3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
B4983-1G
B4983-5G
TCIAB4983-1GEA
39
EUR
TCIAB4983-1G
TCIAB4983-5G
3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one ≥98.0% (por GC)
3-(1,3-Benzodioxol-5-yl)-1-phenyl-2-propen-1-one
Formula:
C₁₆H₁₂O₃ |
Número MDL:
MFCD00022990 Núm. CAS: 644-34-8 |
Resultados test especificaciones
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 120.0 to 124.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...