1-Benzyl-3-phenyl-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Peligro

Sinónimos: 1-Benzyl-3-phenylthiourea

B4997-1G B4997-5G
TCIAB4997-1GEA 59 EUR
TCIAB4997-1G TCIAB4997-5G
1-Benzyl-3-phenyl-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-Benzyl-3-phenyl-2-thiourea
Formula: C₁₄H₁₄N₂S
MW: 242,35 g/mol
Número MDL: MFCD00022120
Núm. CAS: 726-25-0
UN: 2811
ADR: 6.1,III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 153.0 to 157.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR