(2-Bromoethyl)phosphonic acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

B5013-200MG B5013-1G
TCIAB5013-200MGEA 83 EUR
TCIAB5013-200MG TCIAB5013-1G
(2-Bromoethyl)phosphonic acid ≥98.0% (by titrimetric analysis)
(2-Bromoethyl)phosphonic acid
Formula: C₂H₆BrO₃P
MW: 188,94 g/mol
Número MDL: MFCD00013957
Núm. CAS: 999-82-6
UN: 3261
ADR: 8,III

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR