5-Bromo-2-phenylpyridine ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
B5018-1G
B5018-5G
TCIAB5018-1GEA
139
EUR
TCIAB5018-1G
TCIAB5018-5G
5-Bromo-2-phenylpyridine ≥98.0% (por GC)
5-Bromo-2-phenylpyridine
Formula:
C₁₁H₈BrN MW: 234.10 g/mol |
Número MDL:
MFCD04114181 Núm. CAS: 27012-25-5 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...