9,10-Bis[N-(2-naphthyl)anilino]anthracene ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: N,N'-Di(naphthalen-2-yl)-N,N'-diphenylanthracene-9,10-diamine

B5023-200MG B5023-1G
TCIAB5023-200MGEA 131 EUR
TCIAB5023-200MG TCIAB5023-1G
9,10-Bis[N-(2-naphthyl)anilino]anthracene ≥98.0% (by HPLC, total nitrogen)
9,10-Bis[N-(2-naphthyl)anilino]anthracene
Formula: C₄₆H₃₂N₂
MW: 612.78 g/mol
Número MDL: MFCD12022490
Núm. CAS: 473717-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a043546988d64b39cab1d275fc55440d2745d928&vid=a043546988d64b39cab1d275fc55440d2745d928&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR