3'-Bromo-1,4-di(9-carbazolyl)benzene ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
3-Bromo-9-[4-(9H-carbazol-9-yl)phenyl]-9H-carbazole
B5027-200MG
B5027-1G
TCIAB5027-200MGEA
80
EUR
TCIAB5027-200MG
TCIAB5027-1G
3'-Bromo-1,4-di(9-carbazolyl)benzene ≥98.0% (by HPLC, total nitrogen)
3'-Bromo-1,4-di(9-carbazolyl)benzene
Formula:
C₃₀H₁₉BrN₂ MW: 487,40 g/mol |
Número MDL:
MFCD26406219 Núm. CAS: 1537218-76-0 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...