4-Bromo-2,6-diclorofenol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B5034-25G
B5034-5G
TCIAB5034-25GEA
426
EUR
TCIAB5034-25G
TCIAB5034-5G
4-Bromo-2,6-diclorofenol ≥98.0% (by GC, titration analysis)
4-Bromo-2,6-diclorofenol
Formula:
C₆H₃BrCl₂O MW: 241,9 g/mol Pto. Ebullición: 266°C |
Número MDL:
MFCD00019996 Núm. CAS: 3217-15-0 EINECS: 221-740-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | gt. 62.0 and le 66.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...