Bis(4-bromophenyl)diphenylsilane ≥98.0% (por GC)
Proveedor: TCI
B5087-200MG
B5087-1G
TCIAB5087-200MGEA
51
EUR
TCIAB5087-200MG
TCIAB5087-1G
Bis(4-bromophenyl)diphenylsilane ≥98.0% (por GC)
Bis(4-bromophenyl)diphenylsilane
Formula:
C₂₄H₁₈Br₂Si MW: 494,30 g/mol |
Número MDL:
MFCD29089347 Núm. CAS: 18733-91-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...