2,6-Bis(diphenylamino)anthraquinone ≥96.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: 2,6-Bis(diphenylamino)anthracene-9,10-dione

B5092-200MG B5092-1G
TCIAB5092-200MGEA 58 EUR
TCIAB5092-200MG TCIAB5092-1G
2,6-Bis(diphenylamino)anthraquinone ≥96.0% (by HPLC, total nitrogen)
2,6-Bis(diphenylamino)anthraquinone
Formula: C₃₈H₂₆N₂O₂
MW: 542,64 g/mol
Número MDL: MFCD20040460
Núm. CAS: 868850-50-4

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR