N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-di(1-naphthyl)benzidine ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: N1,N1'-([1,1'-Biphenyl]-4,4'-diyl)bis(N1-(naphthalen-1-yl)-N4,N4-diphenylbenzene-1,4-diamine)

B5093-1G B5093-5G
TCIAB5093-1GEA 171 EUR
TCIAB5093-1G TCIAB5093-5G
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-di(1-naphthyl)benzidine ≥98.0% (by HPLC)
N,N'-Bis[4-(diphenylamino)phenyl]-N,N'-di(1-naphthyl)benzidine
Formula: C₆₈H₅₀N₄
MW: 923,18 g/mol
Número MDL: MFCD20488037
Núm. CAS: 910058-11-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d69e2d7675bdfd14139bb42adb8472291e3bf372&vid=d69e2d7675bdfd14139bb42adb8472291e3bf372&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR