tert-Butyl (dimethylphosphono)acetate ≥95.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: tert-Butyl 2-(dimethoxyphosphoryl)acetate, Dimethylphosphonoacetic acid tert-Butyl ester, tert-Butyl dimethylphosphonoacetate, Dimethyl (tert-Butoxycarbonylmethyl)phosphonate, Dimethyl (Boc-methyl)phosphonate

B5094-5G B5094-25G
TCIAB5094-5GEA 28 EUR
TCIAB5094-5G TCIAB5094-25G
tert-Butyl (dimethylphosphono)acetate ≥95.0% (por GC)
tert-Butyl (dimethylphosphono)acetate
Formula: C₈H₁₇O₅P
MW: 224.1913 g/mol
Número MDL: MFCD00042939
Núm. CAS: 62327-21-3

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR