2-(2-Biphenylyl)amino-9,9-dimethylfluorene ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: N-([1,1'-Biphenyl]-2-yl)-9,9-dimethyl-9H-fluoren-2-amine

B5122-1G B5122-5G
TCIAB5122-1GEA 113 EUR
TCIAB5122-1G TCIAB5122-5G
2-(2-Biphenylyl)amino-9,9-dimethylfluorene ≥98.0% (by HPLC)
2-(2-Biphenylyl)amino-9,9-dimethylfluorene
Formula: C₂₇H₂₃N
Número MDL: MFCD19440968
Núm. CAS: 1198395-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3f35e317831bfd09551214e5f709976ef74dd74&vid=c3f35e317831bfd09551214e5f709976ef74dd74&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 118.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR