2-Bromo-6-phenylpyridine ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
B5150-200MG
B5150-1G
TCIAB5150-200MGEA
58
EUR
TCIAB5150-200MG
TCIAB5150-1G
2-Bromo-6-phenylpyridine ≥98.0% (por GC)
2-Bromo-6-phenylpyridine
Formula:
C₁₁H₈BrN MW: 234.1 g/mol |
Número MDL:
MFCD00234691 Núm. CAS: 39774-26-0 |
Resultados test especificaciones
Appearance | White to Yellow to Green powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...