9-(2-Bromoethyl)carbazole ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
B5182-1G
B5182-5G
TCIAB5182-1GEA
138
EUR
TCIAB5182-1G
TCIAB5182-5G
9-(2-Bromoethyl)carbazole ≥98.0% (por GC)
9-(2-Bromoethyl)carbazole
Formula:
C₁₄H₁₂BrN MW: 274,16 g/mol |
Número MDL:
MFCD11127041 Núm. CAS: 75620-93-8 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...