tert-Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: tert-Butylammonium bromide

B5187-1G B5187-5G
TCIAB5187-1GEA 21 EUR
TCIAB5187-1G TCIAB5187-5G
tert-Butylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
tert-Butylamine hydrobromide
Formula: C₄H₁₁N·HBr
MW: 154,05 g/mol
Número MDL: MFCD00674188
Núm. CAS: 60469-70-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR