2-Bromo-3-iodopiridina ≥95.0% (por GC)
Proveedor: TCI
|
Atención
|
B5201-1G
B5201-5G
TCIAB5201-1GEA
47
EUR
TCIAB5201-1G
TCIAB5201-5G
2-Bromo-3-iodopiridina ≥95.0% (por GC)
2-Bromo-3-iodopiridina
Formula:
C₅H₃BrIN MW: 283,89 g/mol |
Número MDL:
MFCD09056782 Núm. CAS: 265981-13-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 96.0 to 100.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...