3-Bromo-4,5-dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

B5252-1G B5252-5G
TCIAB5252-1GEA 141 EUR
TCIAB5252-1G TCIAB5252-5G
3-Bromo-4,5-dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-4,5-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
Número MDL: MFCD00094247
Núm. CAS: 20731-48-0

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 193.0 to 197.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR