Bis(4-fluorophenyl)iodonium trifluoromethanesulphonate ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

B5276-1G B5276-5G
TCIAB5276-1GEA 121 EUR
TCIAB5276-1G TCIAB5276-5G
Bis(4-fluorophenyl)iodonium trifluoromethanesulphonate ≥97.0% (by HPLC, titration analysis)
Bis(4-fluorophenyl)iodonium trifluoromethanesulphonate
Formula: C₁₃H₈F₅IO₃S
Número MDL: MFCD21608479
Núm. CAS: 732306-64-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c29d8f85d2b825d54968f53f0d66aacdca4ff446&vid=c29d8f85d2b825d54968f53f0d66aacdca4ff446&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Ion exchange titration) min. 97.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR