tert-Butyl 4-Formylbenzoate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
tert-Butyl 4-formylbenzoate, 4-Formylbenzoic acid tert-Butyl ester, 4-(tert-Butoxycarbonyl)benzaldehyde
B5414-1G
B5414-5G
TCIAB5414-1GEA
112
EUR
TCIAB5414-1G
TCIAB5414-5G
tert-Butyl 4-Formylbenzoate ≥98.0% (por GC)
tert-Butyl 4-Formylbenzoate
Formula:
C₁₂H₁₄O₃ MW: 206,24 g/mol |
Número MDL:
MFCD01111994 Núm. CAS: 65874-27-3 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...