5H-Benzo[4,5]thieno[3,2-c]carbazole ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

B5434-1G B5434-200MG
TCIAB5434-1GEA 274 EUR
TCIAB5434-1G TCIAB5434-200MG
5H-Benzo[4,5]thieno[3,2-c]carbazole ≥98.0% (by HPLC, total nitrogen)
5H-Benzo[4,5]thieno[3,2-c]carbazole
Formula: C₁₈H₁₁NS
MW: 273,35 g/mol
Número MDL: MFCD22571687
Núm. CAS: 1255308-97-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 215.0 to 219.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR