2-[7-(tert-Butyl)pyren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
Proveedor: TCI
B5489-1G
B5489-5G
TCIAB5489-1GEA
248
EUR
TCIAB5489-1G
TCIAB5489-5G
2-[7-(tert-Butyl)pyren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-[7-(tert-Butyl)pyren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula:
C₂₆H₂₉BO₂ Pto. de fusión: 241°C |
Número MDL:
MFCD31618106 Núm. CAS: 1270030-08-4 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...