2-(4-Bromophenyl)-5-phenylthiophene ≥98.0% (por GC)

Proveedor: TCI

B5491-200MG B5491-1G
TCIAB5491-200MGEA 32 EUR
TCIAB5491-200MG TCIAB5491-1G
2-(4-Bromophenyl)-5-phenylthiophene ≥98.0% (por GC)
2-(4-Bromophenyl)-5-phenylthiophene
Formula: C₁₆H₁₁BrS
MW: 315,23 g/mol
Número MDL: MFCD21648452
Núm. CAS: 118621-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0079de2da5cc8c09404fbd028a5665b6c6fca1a4&vid=0079de2da5cc8c09404fbd028a5665b6c6fca1a4&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 189.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR