Dicyclohexylamine Borane ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Borane - Dicyclohexylamine Complex

B5545-5G B5545-25G
TCIAB5545-5GEA 40 EUR
TCIAB5545-5G TCIAB5545-25G
Dicyclohexylamine Borane ≥95.0% (by titrimetric analysis)
Dicyclohexylamine Borane
Formula: BH₃·C₁₂H₂₃N
MW: 195,16 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 131765-96-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9981fd7a44c728c6cb1260387df61d7decd1701b&vid=9981fd7a44c728c6cb1260387df61d7decd1701b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR