Benzo[b]benzo[4,5]thieno[2,3-d]thiophene ≥97.0% (por GC)
Proveedor: TCI
B5551-200MG
B5551-1G
TCIAB5551-200MGEA
79
EUR
TCIAB5551-200MG
TCIAB5551-1G
Benzo[b]benzo[4,5]thieno[2,3-d]thiophene ≥97.0% (por GC)
Benzo[b]benzo[4,5]thieno[2,3-d]thiophene
Formula:
C₁₄H₈S₂ |
Número MDL:
MFCD02114004 Núm. CAS: 248-70-4 |
|
Resultados test especificaciones
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 218.0 to 222.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...