6-Bromoimidazo[1,2-a]pyrimidine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B5599-1G
B5599-5G
TCIAB5599-1GEA
52
EUR
TCIAB5599-1G
TCIAB5599-5G
6-Bromoimidazo[1,2-a]pyrimidine ≥98.0% (by GC, titration analysis)
6-Bromoimidazo[1,2-a]pyrimidine
Formula:
C₆H₄BrN₃ MW: 198,0200 g/mol |
Número MDL:
MFCD09261434 Núm. CAS: 865156-68-9 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 216.0 to 220.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...