5-Bromo-1,2,3-tris(dodecyloxy)benzene ≥97.0% (por GC)

Proveedor: TCI

B5602-1G B5602-200MG
TCIAB5602-1GEA 239 EUR
TCIAB5602-1G TCIAB5602-200MG
5-Bromo-1,2,3-tris(dodecyloxy)benzene ≥97.0% (por GC)
5-Bromo-1,2,3-tris(dodecyloxy)benzene
Formula: C₄₂H₇₇BrO₃
Número MDL: MFCD32062858
Núm. CAS: 654065-52-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR