N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
N1,N1'-([1,1'-Biphenyl]-4,4'-diyl)bis(N1-phenyl-N4,N4-di-m-tolylbenzene-1,4-diamine), DNTPD
B5634-1G
B5634-5G
TCIAB5634-1GEA
89
EUR
TCIAB5634-1G
TCIAB5634-5G
N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine ≥98.0% (by HPLC)
N,N'-Bis[4-di(m-tolyl)aminophenyl]-N,N'-diphenylbenzidine
Formula:
C₆₄H₅₄N₄ MW: 879,16 g/mol |
Número MDL:
MFCD22372686 Núm. CAS: 199121-98-7 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...