O-Butylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

B5686-1G B5686-5G
TCIAB5686-1GEA 85 EUR
TCIAB5686-1G TCIAB5686-5G
O-Butylhydroxylamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
O-Butylhydroxylamine hydrochloride
Formula: C₄H₁₁NO·HCl
MW: 125,60 g/mol
Número MDL: MFCD00487631
Núm. CAS: 4490-82-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88be6048cc345906d2dabb0f0d2c199f6380b30d&vid=88be6048cc345906d2dabb0f0d2c199f6380b30d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 157.0 to 161.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR