Bis[2-(diisopropylphosphino)ethyl]amine 10% in THF
Proveedor: TCI
|
Peligro
|
B5712-1G
B5712-5G
TCIAB5712-1GEA
109
EUR
TCIAB5712-1G
TCIAB5712-5G
Bis[2-(diisopropylphosphino)ethyl]amine 10% in THF
Bis[2-(diisopropylphosphino)ethyl]amine
Formula:
C₁₆H₃₇NP₂ Pto. de inflamación: -17°C |
Número MDL:
MFCD17018778 Núm. CAS: 131890-26-1 UN: 2056 ADR: 3,II |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid |
Purity (Neutralization titration) | 9.5 to 11.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...