2-Bromo-4,5-difluorobenzoic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B5722-25G
B5722-5G
TCIAB5722-25GEA
196
EUR
TCIAB5722-25G
TCIAB5722-5G
2-Bromo-4,5-difluorobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-4,5-difluorobenzoic acid
Formula:
C₇H₃BrF₂O₂ MW: 237,0000 g/mol |
Número MDL:
MFCD04973422 Núm. CAS: 64695-84-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 118.0 to 122.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...