1-n-Butyl-3-methylimidazolium hexafluoroantimonate ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-Butyl-3-methylimidazolium Hexafluoroantimonate
B5724-5G
B5724-25G
TCIAB5724-5GEA
51
EUR
TCIAB5724-5G
TCIAB5724-25G
1-n-Butyl-3-methylimidazolium hexafluoroantimonate ≥98.0% (by HPLC, total nitrogen)
1-n-Butyl-3-methylimidazolium hexafluoroantimonate
Formula:
C₈H₁₅F₆N₂Sb |
Número MDL:
MFCD07784453 Núm. CAS: 174645-81-9 UN: 3082 ADR: 9,III |
Resultados test especificaciones
Appearance | Colorless to Almost colorless clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...