Tetrafluoroborato de 1-bencil-3-metilimidazolio ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: 1-Benzyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate

B5725-5G B5725-25G
TCIAB5725-5GEA 28 EUR
TCIAB5725-5G TCIAB5725-25G
Tetrafluoroborato de 1-bencil-3-metilimidazolio ≥98.0% (by HPLC, total nitrogen)
Tetrafluoroborato de 1-bencil-3-metilimidazolio
Formula: C₁₁H₁₃BF₄N₂
MW: 260.04 g/mol
Número MDL: MFCD07784449
Núm. CAS: 500996-04-3

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR