Hexafluorofosfato de 1-bencil-3-metilimidazolio ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
B5726-5G
B5726-25G
TCIAB5726-5GEA
32
EUR
TCIAB5726-5G
TCIAB5726-25G
Hexafluorofosfato de 1-bencil-3-metilimidazolio ≥98.0% (by HPLC, total nitrogen)
Hexafluorofosfato de 1-bencil-3-metilimidazolio
Formula:
C₁₁H₁₃F₆N₂P MW: 318.2 g/mol |
Número MDL:
MFCD07784448 Núm. CAS: 433337-11-2 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...