6-Bromo-7-hydroxy-4-(hydroxymethyl)coumarin ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
6-Bromo-7-hydroxy-4-(hydroxymethyl)-2H-chromen-2-one
B5738-250MG
B5738-1G
TCIAB5738-250MGEA
200
EUR
TCIAB5738-250MG
TCIAB5738-1G
6-Bromo-7-hydroxy-4-(hydroxymethyl)coumarin ≥98.0% (by titrimetric analysis)
6-Bromo-7-hydroxy-4-(hydroxymethyl)coumarin
Formula:
C₁₀H₇BrO₄ MW: 271,07 g/mol |
Número MDL:
MFCD25961202 Núm. CAS: 223420-41-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...