1-Butyl-4-methylpyridinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
B5763-1G
B5763-5G
TCIAB5763-1GEA
21
EUR
TCIAB5763-1G
TCIAB5763-5G
1-Butyl-4-methylpyridinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Butyl-4-methylpyridinium bis(trifluoromethanesulphonyl)imide
Formula:
C₁₂H₁₆F₆N₂O₄S₂ MW: 430,38 g/mol Pto. de fusión: 10°C |
Número MDL:
MFCD16038147 Núm. CAS: 475681-62-0 |
Resultados test especificaciones
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...