(7-Bromo-9,9-dimethyl-9H-fluoren-2-yl)boronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 7-Bromo-9,9-dimethyl-9H-fluorene-2-boronic acid

B5773-1G B5773-5G
TCIAB5773-1GEA 115 EUR
TCIAB5773-1G TCIAB5773-5G
(7-Bromo-9,9-dimethyl-9H-fluoren-2-yl)boronic acid (contains varying amounts of Anhydride)
(7-Bromo-9,9-dimethyl-9H-fluoren-2-yl)boronic acid
Formula: C₁₅H₁₄BBrO₂
MW: 316,99 g/mol
Número MDL: MFCD26403351
Núm. CAS: 1213768-48-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 98.0 to 105.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR