2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan ≥96.0% (by HPLC)

Proveedor: TCI

B5822-1G B5822-200MG
TCIAB5822-1GEA 269 EUR
TCIAB5822-1G TCIAB5822-200MG
2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan ≥96.0% (by HPLC)
2,8-Bis(diphenylphosphoryl)dibenzo[b,d]furan
Formula: C₃₆H₂₆O₃P₂
Número MDL: MFCD30607231
Núm. CAS: 911397-27-8

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Melting point 251.0 to 255.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR