5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan ≥98.0% (by GC, titration analysis)
Proveedor: TCI
B5823-1G
B5823-250MG
TCIAB5823-1GEA
131
EUR
TCIAB5823-1G
TCIAB5823-250MG
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan ≥98.0% (by GC, titration analysis)
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[b]furan
Formula:
C₁₄H₁₇BO₃ |
Número MDL:
MFCD04115374 Núm. CAS: 519054-55-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...