Thiophene-2,5-diboronic acid bis(pinacol) ester ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
2,5-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene, Thiophene-2,5-diboronic acid Bis(pinacol) ester, 2,2'-(2,5-Thiophenediyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
B5871-1G
B5871-5G
TCIAB5871-1GEA
45
EUR
TCIAB5871-1G
TCIAB5871-5G
Thiophene-2,5-diboronic acid bis(pinacol) ester ≥98.0% (by GC, titration analysis)
Thiophene-2,5-diboronic acid bis(pinacol) ester
Formula:
C₁₆H₂₆B₂O₄S MW: 336,06 g/mol |
Número MDL:
MFCD09800563 Núm. CAS: 175361-81-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 227.0 to 231.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...